W338210
Ethyl vinyl ketone
≥97%, stabilized, FG
Synonym(s):
1-Penten-3-one, Ethyl vinyl ketone
About This Item
Fragrance grade
Halal
Kosher
meets purity specifications of JECFA
Recommended Products
biological source
synthetic
Quality Level
grade
FG
Fragrance grade
Halal
Kosher
agency
follows IFRA guidelines
meets purity specifications of JECFA
reg. compliance
EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
assay
≥97%
contains
α-tocopherol as stabilizer
refractive index
n20/D 1.419 (lit.)
bp
38 °C/60 mmHg (lit.)
density
0.851 g/mL at 20 °C
0.845 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
documentation
see Safety & Documentation for available documents
food allergen
soybeans
fragrance allergen
no known allergens
organoleptic
garlic; onion; pungent
storage temp.
2-8°C
SMILES string
CCC(=O)C=C
InChI
1S/C5H8O/c1-3-5(6)4-2/h3H,1,4H2,2H3
InChI key
JLIDVCMBCGBIEY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
14.0 °F - closed cup
flash_point_c
-10 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service