Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T6521

Sigma-Aldrich

Sodium tartrate dibasic dihydrate

99.0-101.0% (non-aqueous titration)

Synonym(s):

L-(+)-Tartaric acid disodium salt, Disodium tartrate dihydrate, Sodium tartrate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4Na2O6 · 2H2O
CAS Number:
Molecular Weight:
230.08
Beilstein/REAXYS Number:
6121732
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99.0-101.0% (non-aqueous titration)

SMILES string

[Na+].[Na+].[H]O[H].[H]O[H].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

InChI

1S/C4H6O6.2Na.2H2O/c5-1(3(7)8)2(6)4(9)10;;;;/h1-2,5-6H,(H,7,8)(H,9,10);;;2*1H2/q;2*+1;;/p-2/t1-,2-;;;;/m1..../s1

InChI key

FGJLAJMGHXGFDE-DGFHWNFOSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ai Takemura et al.
Journal of bone and mineral metabolism, 38(5), 631-638 (2020-05-01)
Disuse-induced bone loss is caused by a suppression of osteoblastic bone formation and an increase in osteoclastic bone resorption. There are few data available for the effects of environmental conditions, i.e., atmospheric pressure and/or oxygen concentration, on osteoporosis. This study
Delayed fracture healing and increased callus adiposity in a C57BL/6J murine model of obesity-associated type 2 diabetes mellitus.
Brown ML, Yukata K, Farnsworth CW, et al.
PLoS ONE, 9(6), e99656-e99656 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service