Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E5386

Supelco

Ecgonine methyl ester hydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO3 · HCl · H2O
CAS Number:
Molecular Weight:
253.72
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

O.Cl.COC(=O)[C@H]1[C@@H](O)CC2CCC1N2C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philippe Hantson et al.
Journal of forensic and legal medicine, 18(8), 385-387 (2011-10-25)
Life-threatening complications may occur in body-packers and the rupture of a single packet containing cocaine may lead to fatality. We report the case of a 35-year-old body-packer who developed at the airport clinical signs of cocaine toxicity. There was evidence
Alexander L N van Nuijs et al.
Environmental pollution (Barking, Essex : 1987), 157(1), 123-129 (2008-09-16)
Cocaine abuse, a growing social problem, is currently estimated from population surveys, consumer interviews and crime statistics. A new approach based on the analysis of cocaine (COC) and metabolites, benzoylecgonine (BE) and ecgonine methyl ester (EME), in water samples was
Karl B Scheidweiler et al.
Therapeutic drug monitoring, 32(5), 628-637 (2010-09-04)
Oral fluid is an attractive alternative matrix for drug testing with a noninvasive and directly observed collection, but there are few controlled cocaine administration studies to guide interpretation. While residing on a closed research unit for up to 10 weeks
C Pistos et al.
Rapid communications in mass spectrometry : RCM, 23(23), 3772-3780 (2009-11-11)
At present, no official criteria exist for drug identification using single quadrupole mass spectrometers although the European Union (EU) criteria for compound identification have been adopted. These criteria are evaluated with respect to the confirmation of cocaine and its metabolites
Rosa Cordero et al.
Journal of analytical toxicology, 34(9), 543-548 (2010-11-16)
The concentrations and ratios of cocaine, benzoylecgonine (BE), and ecgoninemethylester (EME) in 360 head hair segments and 34 pubic hair samples collected at coroner's postmortem examinations were reviewed. The cases included diverse histories and causes/circumstances of death. The hair was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service