Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D4882

Supelco

N,N′-Dicyclohexylcarbodiimide–Pentachlorophenol complex

for HPLC derivatization

Synonym(s):

Kovacs Complex

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H25Cl15N2O3
CAS Number:
Molecular Weight:
1005.34
MDL number:
UNSPSC Code:
12000000

grade

for HPLC derivatization

SMILES string

Oc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl.C2CCC(CC2)N=C=NC3CCCCC3

Application

For preparing amino acid pentachlorophenyl esters.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

On the optical purity of peptide active esters prepared by N,N'-dicyclohexylcarbodiimide and "complexes" of N,N'-dicyclohexylcarbodiimide-pentachlorophenol and N,N'-dicyclohexylcarbodiimide-pentafluorophenol.
J Kovacs et al.
Journal of the American Chemical Society, 89(1), 183-184 (1967-01-04)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service