Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C3359

Sigma-Aldrich

6-Carboxy-X-rhodamine N-succinimidyl ester

Synonym(s):

6-ROX, SE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H33N3O7
CAS Number:
Molecular Weight:
631.67
MDL number:
UNSPSC Code:
12171500

fluorescence

λex ~570 nm; λem ~595 nm in 0.1 M Tris pH 8.0

shipped in

wet ice

storage temp.

−20°C

Application

Succinimidyl ester derivative of carboxy-X-rhodamine, widely used for oligonucleotide labeling and automated DNA sequencing applications.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service