95447
Bis[tris(4-(1H,1H,2H,2H-perfluorodecyl)phenyl)phosphine]palladium(II) dichloride
technical, ≥80% (AAS)
Synonym(s):
Bis[tris(4-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)phenyl)phosphine]palladium(II) dichloride
Select a Size
About This Item
Recommended Products
grade
technical
Quality Level
assay
≥80% (AAS)
form
powder
reaction suitability
reagent type: catalyst
core: palladium
mp
175-180 °C
SMILES string
Cl[Pd]Cl.FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCc1ccc(cc1)P(c2ccc(CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)cc2)c3ccc(CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)cc3.FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCc4ccc(cc4)P(c5ccc(CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)cc5)c6ccc(CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)cc6
InChI
1S/2C48H24F51P.2ClH.Pd/c2*49-25(50,28(55,56)31(61,62)34(67,68)37(73,74)40(79,80)43(85,86)46(91,92)93)16-13-19-1-7-22(8-2-19)100(23-9-3-20(4-10-23)14-17-26(51,52)29(57,58)32(63,64)35(69,70)38(75,76)41(81,82)44(87,88)47(94,95)96)24-11-5-21(6-12-24)15-18-27(53,54)30(59,60)33(65,66)36(71,72)39(77,78)42(83,84)45(89,90)48(97,98)99;;;/h2*1-12H,13-18H2;2*1H;/q;;;;+2/p-2
InChI key
KONKDVXTNQEYIF-UHFFFAOYSA-L
Related Categories
General description
Application
Packaging
signalword
Danger
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1
target_organs
Liver
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service