93349
Trizma® base
BioUltra, ≥99.8% (T)
Synonym(s):
2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol
About This Item
Recommended Products
description
aminopeptidase substrate
product line
BioUltra
assay
≥99.8% (T)
impurities
insoluble matter, passes filter test
ign. residue
≤0.01% (as SO4)
loss
≤0.5% loss on drying, 110 °C
pH
10.5-12.0 (25 °C, 4 M in H2O)
useful pH range
7-9
pKa (25 °C)
8.1
bp
219-220 °C/10 mmHg (lit.)
mp
167-172 °C (lit.)
168-172 °C
solubility
H2O: 4 M at 20 °C, clear, colorless
anion traces
chloride (Cl-): ≤5 mg/kg
sulfate (SO42-): ≤5 mg/kg
cation traces
Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg
λ
4 M in H2O
UV absorption
λ: 260 nm Amax: 0.10
λ: 280 nm Amax: 0.08
SMILES string
NC(CO)(CO)CO
InChI
1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2
InChI key
LENZDBCJOHFCAS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service