Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92793

Sigma-Aldrich

1,5,9-Trimethyl-1,5,9-triazacyclododecane

technical, ≥90% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H27N3
CAS Number:
Molecular Weight:
213.36
Beilstein/REAXYS Number:
4858723
MDL number:
UNSPSC Code:
12350000
PubChem Substance ID:

grade

technical

assay

≥90% (GC)

refractive index

n20/D 1.483 (lit.)

bp

58-60 °C/0.1 mmHg (lit.)

density

0.91 g/mL at 20 °C (lit.)

SMILES string

CN1CCCN(C)CCCN(C)CCC1

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service