92677
Di-μ-hydroxo-bis(N,N,N′,N′-tetramethylethylenediamine)copper(II) chloride
purum, ≥97.0% (RT)
Synonym(s):
Chlorohydroxo(N,N,N′,N′-tetrametylethylenediamine)copper(II), Copper(II) chloride hydroxide (N,N,N′,N′-tetrametylethylenediamine) Complex Dimer, Cu-TMEDA catalyst
About This Item
grade
purum
assay
≥97.0% (RT)
mp
138-148 °C
SMILES string
O[Cu]Cl.O[Cu]Cl.CN(C)CCN(C)C.CN(C)CCN(C)C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service