Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92415

Sigma-Aldrich

3,5,5-Trimethyl-2-cyclohexen-1-ol

technical, ≥95% (GC)

Synonym(s):

Isophorol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H6OH
CAS Number:
Molecular Weight:
140.22
Beilstein/REAXYS Number:
1929064
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

assay

≥95% (GC)

refractive index

n20/D 1.472 (lit.)
n20/D 1.472

bp

79.5-81.5 °C/8 mmHg (lit.)

density

0.918 g/mL at 25 °C (lit.)

SMILES string

CC1=CC(O)CC(C)(C)C1

InChI

1S/C9H16O/c1-7-4-8(10)6-9(2,3)5-7/h4,8,10H,5-6H2,1-3H3

InChI key

LDRWAWZXDDBHTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service