Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91956

Sigma-Aldrich

4′,6,7-Trihydroxyisoflavone

≥96.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
Beilstein/REAXYS Number:
1291621
MDL number:
UNSPSC Code:
12352200

assay

≥96.0% (TLC)

SMILES string

C1=CC(=CC=C1C2=COC3=CC(=C(C=C3C2=O)O)O)O

InChI

InChI=1S/C15H10O5/c16-9-3-1-8(2-4-9)11-7-20-14-6-13(18)12(17)5-10(14)15(11)19/h1-7,16-18H

InChI key

GYLUFQJZYAJQDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service