Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91668

Sigma-Aldrich

Trifluoroacetaldehyde ethyl hemiacetal

technical

Synonym(s):

1-Ethoxy-2,2,2-trifluoroethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CH(OH)OCH2CH3
CAS Number:
Molecular Weight:
144.09
Beilstein/REAXYS Number:
906797
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

refractive index

n20/D 1.342 (lit.)
n20/D 1.342

bp

104-105 °C/745 mmHg (lit.)

density

1.221 g/mL at 25 °C (lit.)

SMILES string

[Na+].[O-]c1ccccc1

InChI

1S/C4H7F3O2/c1-2-9-3(8)4(5,6)7/h3,8H,2H2,1H3

InChI key

KLXJPQNHFFMLIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service