90245
2,3,4-Tri-O-acetyl-α-D-arabinopyranosyl isothiocyanate
for chiral derivatization, LiChropur™, ≥98.0%
Synonym(s):
AITC
About This Item
grade
derivatization grade ((chiral))
for chiral derivatization
product line
ChiraSelect™
assay
≥98.0% (sum of enantiomers, HPLC)
≥98.0%
optical activity
[α]20/D +50±1°, c = 1% in chloroform stab. with amylenes
optical purity
enantiomeric ratio: ≥99.5:0.5 (HPLC)
quality
LiChropur™
storage temp.
−20°C
SMILES string
CC(=O)O[C@@H]1CO[C@H](N=C=S)[C@@H](OC(C)=O)[C@@H]1OC(C)=O
InChI
1S/C12H15NO7S/c1-6(14)18-9-4-17-12(13-5-21)11(20-8(3)16)10(9)19-7(2)15/h9-12H,4H2,1-3H3/t9-,10-,11+,12+/m1/s1
InChI key
RCHZRAFPHKCWRI-WYUUTHIRSA-N
Packaging
Other Notes
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service