Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901984

Sigma-Aldrich

PC biotin-alkyne

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H52N6O12S
CAS Number:
Molecular Weight:
780.89
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

form

liquid

availability

available only in USA

storage temp.

−20°C

Application

PC biotin-alkyne is an azide-reactive photoclevable biotin probe that allows for reagent-free release of the captured biomolecules from streptavidin. This reagent contains a biotin moiety linked to an alkyne group through a spacer arm containing a photocleavable moiety. Captured biomolecules can be efficiently photoreleased, typically >90% in 5-25 minutes using an inexpensive, near-UV, low-intensity lamp (e.g. 365 nm lamp at 1-5 mW/cm2).

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Self-react. C

Storage Class

5.2 - Organic peroxides and self-reacting hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service