Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90135

Sigma-Aldrich

p-Tolyl [2-(trimethylsilyl)ethynyl] sulfone

purum, ≥97.0% (CHN)

Synonym(s):

1-(p-Toluenesulfonyl)-2-(trimethylsilyl)acetylene, 2-(Trimethylsilyl)ethynyl p-tolyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiC≡CSO2C6H4CH3
CAS Number:
Molecular Weight:
252.40
Beilstein/REAXYS Number:
2650368
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥97.0% (CHN)

mp

80-83 °C (lit.)

SMILES string

Cc1ccc(cc1)S(=O)(=O)C#C[Si](C)(C)C

InChI

1S/C12H16O2SSi/c1-11-5-7-12(8-6-11)15(13,14)9-10-16(2,3)4/h5-8H,1-4H3

InChI key

MMUNNGUGQXEUBR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Powerful Michael acceptor for vinylations[1][2]; Sterically hindered dipolarophile for cycloadditions[3][4]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A. Padwa et al.
Tetrahedron, 46, 1145-1145 (1990)
Tetrahedron Letters, 27, 219-219 (1986)
A. Padwa et al.
The Journal of Organic Chemistry, 52, 4760-4760 (1987)
T. Ohnuma et al.
The Journal of Organic Chemistry, 47, 4713-4713 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service