Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89509

Millipore

Heptakis-(2,6-di-O-ethyl)-β-cyclodextrin

suitable for GC

Synonym(s):

2,6-Di-O-ethyl-β-cyclodextrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C70H126O35
CAS Number:
Molecular Weight:
1527.73
Beilstein/REAXYS Number:
8399169
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

extent of labeling

~14 substitution (ethyl groups)

technique(s)

gas chromatography (GC): suitable

mp

~215 °C (dec.)

solubility

DMSO: soluble
chloroform: soluble

SMILES string

CCOC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](OCC)[C@H](O[C@@H]3COCC)O[C@H]4[C@H](O)[C@@H](OCC)[C@H](O[C@@H]4COCC)O[C@H]5[C@H](O)[C@@H](OCC)[C@H](O[C@@H]5COCC)O[C@H]6[C@H](O)[C@@H](OCC)[C@H](O[C@@H]6COCC)O[C@H]7[C@H](O)[C@@H](OCC)[C@H](O[C@@H]7COCC)O[C@H]8[C@H](O)[C@@H](OCC)[C@H](O[C@@H]8COCC)O[C@H]1[C@H](O)[C@H]2OCC

Other Notes

Preparation of a β-cyclodextrin-based fluorosensor for tetracyclines[1]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Hirayama et al.
Pharmaceutical research, 10(2), 208-213 (1993-02-01)
Heptakis(2,6-di-O-ethyl)-beta-cyclodextrin (DE-beta-CyD) was prepared and its 1H and 13C nuclear magnetic resonance (NMR) signals in DMSO-d6 were unequivocally assigned by two-dimensional COSY and ROESY. The results on 1H coupling constants indicated that all ethylated glucose units are in a 4C1
F Hirayama et al.
Journal of pharmaceutical sciences, 77(3), 233-236 (1988-03-01)
Heptakis(2,6-di-O-ethyl)-beta-cyclodextrin (2) was prepared and its physicochemical properties, such as aqueous solubility and surface activity, were compared with those of beta-cyclodextrin (1) and heptakis(2,6-di-O-methyl)-beta-cyclodextrin (3). A possible utility of 2 as a sustained-release drug carrier was examined using a water-soluble
Z. Gong et al.
Analytica Chimica Acta, 351, 205-205 (1997)
M Umemura et al.
Drug design and delivery, 6(4), 297-310 (1990-10-01)
The complex-forming abilities of 2,6-di-O-ethyl-beta-cyclodextrin (DE-beta-CD), and its effect on the release of nitroglycerin (TNG) from formulations of the compound, were studied and compared with corresponding properties of beta-cyclodextrin (beta-CD) and 2,6-di-O-methyl-beta-cyclodextrin (DM-beta-CD). Complex formation was confirmed by differential scanning
K Harata et al.
Carbohydrate research, 329(3), 597-607 (2000-12-29)
Heptakis(2,6-di-O-ethyl)-beta-cyclodextrin (DE-beta-CD) was crystallized in two forms from hexane and 95% aqueous methanol, respectively: A form I crystal with the space group P2(1)2(1)2(1) and a form II crystal with the space group P3(1). In both crystals, DE-beta-CD molecules are in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service