88935
N-Thionylaniline
purum, ≥97.0% (GC)
Synonym(s):
N-Sulfinyl-aniline
About This Item
grade
purum
assay
≥97.0% (GC)
color
very strongly colored
refractive index
n20/D 1.627 (lit.)
bp
200 °C (lit.)
83 °C/15 mmHg (lit.)
density
1.236 g/mL at 25 °C (lit.)
SMILES string
O=S=Nc1ccccc1
InChI
1S/C6H5NOS/c8-9-7-6-4-2-1-3-5-6/h1-5H
InChI key
FIOJWGRGPONADF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
replaced by
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service