Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

88340

Sigma-Aldrich

Thiamine monophosphate chloride dihydrate

≥99.0% (HPLC)

Synonym(s):

Aneurine monophosphate chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18ClN4O4PS · 2H2O
CAS Number:
Molecular Weight:
416.82
Beilstein/REAXYS Number:
3844977
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥99.0% (HPLC)

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

storage temp.

2-8°C

SMILES string

[Cl-].Cc1ncc(C[n+]2csc(CCOP(O)(O)=O)c2C)c(N)n1

InChI

1S/C12H17N4O4PS.ClH/c1-8-11(3-4-20-21(17,18)19)22-7-16(8)6-10-5-14-9(2)15-12(10)13;/h5,7H,3-4,6H2,1-2H3,(H3-,13,14,15,17,18,19);1H

InChI key

GUGWNSHJDUEHNJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Thiamine: Twenty Years of Progress

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annals of the New York Academy of Sciences, 378, 468-468 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service