Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

87202

Sigma-Aldrich

Tetradecylamine

puriss., ≥98.5% (GC)

Synonym(s):

1-Aminotetradecane, Myristylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13NH2
CAS Number:
Molecular Weight:
213.40
Beilstein/REAXYS Number:
1633740
EC Number:
MDL number:
UNSPSC Code:
12352100

grade

puriss.

assay

≥98.5% (GC)

bp

162 °C/15 mmHg (lit.)

mp

38-40 °C (lit.)
38-40 °C

SMILES string

CCCCCCCCCCCCCCN

InChI

1S/C14H31N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15/h2-15H2,1H3

InChI key

PLZVEHJLHYMBBY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Salem et al.
Journal of clinical periodontology, 14(1), 44-47 (1987-01-01)
The surfactants tetradecylamine, hexadecylamine and chlorhexidine have been compared with regard to their ability to inhibit microbial growth. Antibacterial activity was assessed by tube dilution methods. Tetradecylamine and chlorhexidine were similar in antibacterial activity, being effective at low concentrations against
Takashi Kitahara et al.
International journal of antimicrobial agents, 27(1), 51-57 (2005-12-02)
The objective of this study was to investigate the in vitro activities of lauric acid and myristylamine in combination with six antimicrobial agents against methicillin-resistant Staphylococcus aureus (MRSA). The combination effect of lipids and antimicrobial agents was evaluated by the
Y Murata et al.
Caries research, 24(2), 113-116 (1990-01-01)
Some antimicrobial compounds were tested for their antiplaque activity by a simple method involving measurement of the weight of Streptococcus sobrinus plaque on a glass surface. Chlorhexidine, cetylpyridinium chloride, and n-tetradecylamine reduced the weight of plaque produced by S. sobrinus
Phuong D Nguyen et al.
FEMS microbiology ecology, 66(1), 136-142 (2008-08-30)
The anaerobic degradation of tetradecylamine and other long-chain alkylamines by a newly isolated denitrifying bacterium was studied. Strain ZN6 was isolated from a mixture of soil and active sludge and was identified as representing Pseudomonas stutzeri, based on partial 16S
T Ohtsuka et al.
Archives of biochemistry and biophysics, 280(1), 74-79 (1990-07-01)
NADPH oxidase activity in a membrane fraction prepared from phorbol 12-myristate 13-acetate (PMA)-stimulated guinea pig polymorphonuclear leukocytes (PMNL) was inhibited by positively charged myristylamine. The inhibitory effect of myristylamine was significantly suppressed by simultaneous addition of a negatively charged fatty

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service