Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

86733

Sigma-Aldrich

1,4,8,11-Tetraazacyclotetradecane-5,7-dione

purum, ≥97.0% (CHN)

Synonym(s):

Dioxocyclam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20N4O2
CAS Number:
Molecular Weight:
228.29
Beilstein/REAXYS Number:
1214926
MDL number:
UNSPSC Code:
12162002

grade

purum

assay

≥97.0% (CHN)

mp

172-176 °C (lit.)
172-176 °C

SMILES string

O=C1CC(=O)NCCNCCCNCCN1

InChI

1S/C10H20N4O2/c15-9-8-10(16)14-7-5-12-3-1-2-11-4-6-13-9/h11-12H,1-8H2,(H,13,15)(H,14,16)

Inchi Key

BGVLBVASHIQNIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Ligand for the selective complexation of transition metals[1][2][3]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L. Fabbrizzi et al.
Inorg. Synth., 23, 82-82 (1985)
E. Kimura et al.
Journal of the Chemical Society. Chemical Communications, 1020-1020 (1986)
A. Jyo et al.
Analytical Sciences, 8, 195-195 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service