85905
Stuart™ Ringertz Medium
suitable for microbiology
Synonym(s):
Transport Medium according to Stuart™, modified by Ringertz
About This Item
Recommended Products
shelf life
limited shelf life, expiry date on the label
composition
agar, 8 g/L
calcium chloride, 0.1 g/L
methylene blue, 0.002 g/L
sodium glycerophosphate, 10 g/L
sodium thioglycolate, 1 g/L
final pH
7.4±0.2 (37 °C)
application(s)
microbiology
Application
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service