83911
Ristomycin monosulfate
crystallized, ≥99.0% (TLC)
Synonym(s):
Ristocetin sulfate salt
About This Item
Recommended Products
assay
≥99.0% (TLC)
quality
crystallized
solubility
H2O: 20 mg/mL, clear, colorless to light yellow
mode of action
cell wall synthesis | interferes
storage temp.
2-8°C
SMILES string
COC(=O)[C@@H]1NC(=O)[C@H]2NC(=O)C(NC(=O)[C@@H]3NC(=O)[C@H]4NC(=O)[C@H](NC(=O)[C@H](N)c5ccc(O)c(Oc6cc4cc(O)c6C)c5)[C@H](O)c7ccc(Oc8cc3cc(Oc9ccc(cc9)[C@H]2O[C@H]%10C[C@@H](N)[C@@H](O)[C@H](C)O%10)c8O[C@@H]%11O[C@H](CO[C@@H]%12O[C@@H](C)[C@H](O)[C@@H](O)[C@H]%12O)[C@@H](O)[C@H](O)[C@H]%11O[C@H]%13O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]%13O[C@@H]%14OC[C@@H](O)[C@@H](O)[C@@H]%14O)cc7)c%15ccc(O)c(c%15)-c%16c(O[C@@H]%17O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]%17O)cc(O)cc1%16
InChI
1S/C95H110N8O44/c1-30-47(109)18-37-20-49(30)139-50-19-35(9-16-46(50)108)59(97)84(125)102-64-68(113)33-5-11-40(12-6-33)137-52-21-38-22-53(81(52)145-95-83(76(121)72(117)56(143-95)29-134-91-78(123)73(118)67(112)32(3)136-91)147-94-82(75(120)71(116)55(27-105)142-94)146-92-77(122)69(114)48(110)28-133-92)138-41-13-7-34(8-14-41)80(144-57-25-44(96)66(111)31(2)135-57)65-89(130)101-63(90(131)132-4)43-23-39(106)24-51(140-93-79(124)74(119)70(115)54(26-104)141-93)58(43)42-17-36(10-15-45(42)107)60(85(126)103-65)98-87(128)62(38)99-86(127)61(37)100-88(64)129/h5-24,31-32,44,48,54-57,59-80,82-83,91-95,104-124H,25-29,96-97H2,1-4H3,(H,98,128)(H,99,127)(H,100,129)(H,101,130)(H,102,125)(H,103,126)/t31-,32-,44+,48+,54+,55+,56+,57-,59+,60?,61-,62+,63?,64+,65-,66-,67-,68+,69+,70+,71+,72+,73+,74-,75-,76-,77-,78+,79-,80+,82-,83+,91+,92-,93+,94+,95-/m0/s1
InChI key
VUOPFFVAZTUEGW-ZNAZZOOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
replaced by
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service