Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

79134BC

Millipore

GDS variable Amp Tube Holder, 72 well

BioControl Systems, for use with GDS Rotor-Gene®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104017
NACRES:
NA.85

manufacturer/tradename

BioControl Systems

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS Rotor-Gene®

storage temp.

room temp

Legal Information

Rotor-Gene is a registered trademark of Qiagen GmbH

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service