79010
3-Phenylpropylamine
purum, ≥98.0% (GC)
Synonym(s):
γ-Aminopropylbenzene
About This Item
Recommended Products
grade
purum
assay
≥98.0% (GC)
refractive index
n20/D 1.524 (lit.)
n20/D 1.526
bp
221 °C (lit.)
density
0.951 g/mL at 25 °C (lit.)
SMILES string
NCCCc1ccccc1
InChI
1S/C9H13N/c10-8-4-7-9-5-2-1-3-6-9/h1-3,5-6H,4,7-8,10H2
InChI key
LYUQWQRTDLVQGA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
Hazard Classifications
Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
195.8 °F - closed cup
flash_point_c
91 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service