Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78865

Sigma-Aldrich

N-Phenyl-1-naphthylamine

technical, ≥95% (N)

Synonym(s):

1-(N-phenylamino)naphthalene, N-(1-Naphthyl)aniline, NPN

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7NHC6H5
CAS Number:
Molecular Weight:
219.28
Beilstein/REAXYS Number:
2211174
EC Number:
MDL number:
UNSPSC Code:
12190000
Pricing and availability is not currently available.

grade

technical

assay

≥95% (N)

bp

226 °C/15 mmHg (lit.)

mp

60-62 °C (lit.)

SMILES string

N(c1ccccc1)c2cccc3ccccc23

InChI

1S/C16H13N/c1-2-9-14(10-3-1)17-16-12-6-8-13-7-4-5-11-15(13)16/h1-12,17H

InChI key

XQVWYOYUZDUNRW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Kröber et al.
Insect biochemistry and molecular biology, 96, 36-50 (2018-04-16)
There is increasing interest in the development of effective mosquito repellents of natural origin to reduce transmission of diseases such as malaria and yellow fever. To achieve this we have employed an in vitro competition assay involving odorant-binding proteins (OBPs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service