Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

76167

Sigma-Aldrich

Palmitic acid N-hydroxysuccinimide ester

technical, ≥90% (HPLC)

Synonym(s):

N-(Palmitoyloxy)succinimide, N-Succinimidyl palmitate, Palmitic acid N-succinimidyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H35NO4
CAS Number:
Molecular Weight:
353.50
Beilstein/REAXYS Number:
1547411
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
Pricing and availability is not currently available.

grade

technical

assay

≥90% (HPLC)

mp

86-91 °C

SMILES string

CCCCCCCCCCCCCCCC(=O)ON1C(=O)CCC1=O

InChI

1S/C20H35NO4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-20(24)25-21-18(22)16-17-19(21)23/h2-17H2,1H3

InChI key

OTNHQVHEZCBZQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service