Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74960

Sigma-Aldrich

Trimethoxy(7-octen-1-yl)silane

technical, ≥85% (GC)

Synonym(s):

(7-Octen-1-yl)trimethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CH(CH2)6Si(OCH3)3
CAS Number:
Molecular Weight:
232.39
Beilstein/REAXYS Number:
2437131
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

technical

assay

≥85% (GC)

refractive index

n20/D 1.427 (lit.)
n20/D 1.427

bp

48-49 °C/0.1 mmHg (lit.)

density

0.928 g/mL at 25 °C (lit.)

SMILES string

CO[Si](CCCCCCC=C)(OC)OC

InChI

1S/C11H24O3Si/c1-5-6-7-8-9-10-11-15(12-2,13-3)14-4/h5H,1,6-11H2,2-4H3

InChI key

RKLXSINPXIQKIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

201.2 °F - closed cup

flash_point_c

94 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service