Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73952

Supelco

4-Nitrososulfamethoxazole

analytical standard

Synonym(s):

N-(5-Methyl-3-isoxazolyl)-4-nitrosobenzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9N3O4S
CAS Number:
Molecular Weight:
267.26
Beilstein/REAXYS Number:
6338308
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤2.0% water (calc. from elemental analysis)

application(s)

pharmaceutical

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC1=CC(NS(=O)(C2=CC=C(N=O)C=C2)=O)=NO1

InChI

1S/C10H9N3O4S/c1-7-6-10(12-17-7)13-18(15,16)9-4-2-8(11-14)3-5-9/h2-6H,1H3,(H,12,13)

InChI key

GHNQGDUYHCZZPT-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ki-Jun Yoon et al.
Cell, 171(4), 877-889 (2017-10-03)
N6-methyladenosine (m6A), installed by the Mettl3/Mettl14 methyltransferase complex, is the most prevalent internal mRNA modification. Whether m6A regulates mammalian brain development is unknown. Here, we show that m6A depletion by Mettl14 knockout in embryonic mouse brains prolongs the cell cycle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service