Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73782

Sigma-Aldrich

4-Nitrophthalic anhydride

technical, ~90% (T)

Synonym(s):

4-Nitrophthalic acid anhydride, 5-Nitroisobenzofuran-1,3-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H3NO5
CAS Number:
Molecular Weight:
193.11
Beilstein/REAXYS Number:
179682
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

grade

technical

assay

~90% (T)

mp

116-120 °C (lit.)
116-120 °C

SMILES string

[O-][N+](=O)c1ccc2C(=O)OC(=O)c2c1

InChI

1S/C8H3NO5/c10-7-5-2-1-4(9(12)13)3-6(5)8(11)14-7/h1-3H

InChI key

MMVIDXVHQANYAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service