73040
4-Nitrobenzenesulfonyl chloride
purum, ≥95.0% (AT), yellow
About This Item
grade
purum
assay
≥95.0% (AT)
color
yellow
mp
66-70 °C (lit.)
75-79 °C
SMILES string
[O-][N+](=O)c1ccc(cc1)S(Cl)(=O)=O
InChI
1S/C6H4ClNO4S/c7-13(11,12)6-3-1-5(2-4-6)8(9)10/h1-4H
InChI key
JXRGUPLJCCDGKG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Repr. 2 - Skin Sens. 1A
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service