72650
1-Ethyl-4-nitrobenzene
purum, ≥99.0% (GC)
Synonym(s):
4-Nitroethylbenzene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H9NO2
CAS Number:
Molecular Weight:
151.16
Beilstein/REAXYS Number:
1862416
EC Number:
MDL number:
UNSPSC Code:
12352103
grade
purum
assay
≥99.0% (GC)
refractive index
n20/D 1.545
bp
250-253 °C
mp
−11-−8 °C
SMILES string
CCc1ccc(cc1)[N+]([O-])=O
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service