Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

68704

Sigma-Aldrich

(2S,3S)-trans-2-Methyl-3-phenyloxirane-2-methanol

purum, ≥95.0% (sum of enantiomers, GC)

Synonym(s):

(2S,3S)-(−)-trans-2-Methyl-3-phenylglycidol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
Beilstein/REAXYS Number:
5862068
MDL number:
UNSPSC Code:
12352000

grade

purum

assay

≥95.0% (sum of enantiomers, GC)

optical activity

[α]20/D −16±2°, c = 2% in chloroform

mp

~55 °C

shipped in

wet ice

storage temp.

−20°C

SMILES string

C[C@@]1(CO)O[C@H]1c2ccccc2

Application

Chiral building block

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service