Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

68260

Sigma-Aldrich

(±)-3-Methyl-2-oxovaleric acid sodium salt

purum, ≥99.0% (NT)

Synonym(s):

(±)-Sodium 3-methyl-2-oxovalerate, 3-Methyl-2-oxopentanoic acid sodium salt, DL-α-Keto-β-methylvaleric acid sodium salt, Ketoisoleucine sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)COCOONa
CAS Number:
Molecular Weight:
152.12
Beilstein/REAXYS Number:
7779663
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

grade

purum

assay

≥99.0% (NT)

mp

196-199 °C

SMILES string

[Na+].CCC(C)C(=O)C([O-])=O

InChI

1S/C6H10O3.Na/c1-3-4(2)5(7)6(8)9;/h4H,3H2,1-2H3,(H,8,9);/q;+1/p-1

InChI key

SMDJDLCNOXJGKC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service