Skip to Content
MilliporeSigma
All Photos(1)

Documents

66955

Supelco

Copper (impurities)

manufactured by MBH Analytical Ltd (39X 17868)

Synonym(s):

Copper

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cu
CAS Number:
Molecular Weight:
63.55
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

material

disks

grade

analytical standard

resistivity

1.673 μΩ-cm, 20°C

technique(s)

XRF: suitable

diam. × H

40 mm × 17 mm , dimensions are approximate

bp

2567 °C (lit.)

mp

1083.4 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

format

matrix material

SMILES string

[Cu]

InChI

1S/Cu

InChI key

RYGMFSIKBFXOCR-UHFFFAOYSA-N

Analysis Note

Values of analytes vary lot to lot.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marius Andruh
Chimia, 67(6), 383-387 (2013-08-16)
The node-and-spacer approach is widely employed in crystal engineering to construct coordination polymers. It consists of self-assembly processes involving mononuclear cationic species and exo-dentate ligands. We enlarged this strategy using preformed homo- and heterometallic complexes as nodes. The presence of
Kessarin Thanapirom et al.
Journal of the Medical Association of Thailand = Chotmaihet thangphaet, 96(2), 150-156 (2013-08-14)
Wilson's disease (WD) is an inherited disorder of copper metabolism, leading to hepatic and neuropsychiatric manifestations. The long-term outcome of patients with Wilson's disease presenting with acute liver failure (ALF) versus those with acute-on-chronic liver failure (ACLF) has not been
Taekyung Yu et al.
Journal of nanoscience and nanotechnology, 13(5), 3250-3253 (2013-07-19)
Copper (Cu) nanoparticles with controllable sizes were successfully synthesized by using a modified reverse micelle method, in which copper(II) acetate was reacted with L-ascorbic acid in a solution containing water and xylene in the presence of oleic acid and oleylamine
Xuejiao Tong et al.
Journal of environmental sciences (China), 25(4), 652-658 (2013-08-09)
The removal efficiency of copper (Cu(II)) from an actual acidic electroplating effluent by biochars generated from canola, rice, soybean and peanut straws was investigated. The biochars simultaneously removed Cu(II) from the effluent, mainly through the mechanisms of adsorption and precipitation
Ping Ning et al.
Journal of environmental sciences (China), 25(4), 808-814 (2013-08-09)
Metal (Cu, Co, or Zn) loaded ZSM-5 and Y zeolite adsorbents were prepared for the adsorption of hydrogen cyanide (HCN) toxic gas. The results showed that the HCN breakthrough capacity was enhanced significantly when zeolites were loaded with Cu. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service