634042
Promethazine-d4 hydrochloride
98 atom % D, 98% (CP)
Synonym(s):
Phenothiazine-1,3,7,9-d4 hydrochloride
About This Item
isotopic purity
98 atom % D
assay
98% (CP)
mp
212-216 °C (lit.)
mass shift
M+4
SMILES string
Cl.[2H]c1cc([2H])c2N(CC(C)N(C)C)c3c([2H])cc([2H])cc3Sc2c1
InChI
1S/C17H20N2S.ClH/c1-13(18(2)3)12-19-14-8-4-6-10-16(14)20-17-11-7-5-9-15(17)19;/h4-11,13H,12H2,1-3H3;1H/i6D,7D,8D,9D;
InChI key
XXPDBLUZJRXNNZ-RPMJCVFWSA-N
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service