Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

62546

Sigma-Aldrich

Lithium isopropoxide solution

technical, ~2 M in THF

Synonym(s):

Lithium 2-propanolate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOLi
CAS Number:
Molecular Weight:
66.03
Beilstein/REAXYS Number:
3618614
MDL number:
UNSPSC Code:
12352300

grade

technical

concentration

~2 M in THF

density

0.897 g/mL at 20 °C

SMILES string

[Li+].CC(C)[O-]

InChI

1S/C3H7O.Li/c1-3(2)4;/h3H,1-2H3;/q-1;+1

InChI key

HAUKUGBTJXWQMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service