61695
17α(H)-22,29,30-Trisnorhopan solution
0.1 mg/mL in isooctane, analytical standard
Synonym(s):
17α(H)-22,29,30-Trisnor-A′-neogammacerane
About This Item
Recommended Products
grade
analytical standard
Quality Level
shelf life
limited shelf life, expiry date on the label
concentration
0.1 mg/mL in isooctane
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
food and beverages
format
single component solution
storage temp.
−20°C
SMILES string
[H][C@@]12CCC[C@]1(C)C3CCC4[C@@]5(C)CCCC(C)(C)C5CC[C@@]4(C)[C@]3(C)CC2
InChI
1S/C27H46/c1-23(2)14-8-16-25(4)20(23)13-18-27(6)22(25)11-10-21-24(3)15-7-9-19(24)12-17-26(21,27)5/h19-22H,7-18H2,1-6H3
InChI key
WPKYQECPNNWDJY-UHFFFAOYSA-N
Application
signalword
Danger
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Central nervous system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
10.4 °F - closed cup
flash_point_c
-12 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
We provide high-quality, relevant hopane reference standards to be used as biomarkers for petrochemical oil field remediation and spill analysis.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service