Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

60401

Supelco

Potassium ionophore II

Selectophore, function tested

Synonym(s):

BB15C5, Bis[(benzo-15-crown-5)-4′-ylmethyl] pimelate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H52O14
CAS Number:
Molecular Weight:
720.80
Beilstein/REAXYS Number:
1676229
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

quality

function tested

SMILES string

O=C(CCCCCC(=O)OCc1ccc2OCCOCCOCCOCCOc2c1)OCc3ccc4OCCOCCOCCOCCOc4c3

InChI

1S/C37H52O14/c38-36(50-28-30-6-8-32-34(26-30)48-24-20-44-16-12-40-10-14-42-18-22-46-32)4-2-1-3-5-37(39)51-29-31-7-9-33-35(27-31)49-25-21-45-17-13-41-11-15-43-19-23-47-33/h6-9,26-27H,1-5,10-25,28-29H2

InChI key

LTZRCLYZVSXCTC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Potassium ionophore for assaying K+-activities[1][2]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah Sierra Smaga et al.
Structure (London, England : 1993), 27(8), 1234-1245 (2019-06-04)
The human antiviral protein MxB is a restriction factor that fights HIV infection. Previous experiments have demonstrated that MxB targets the HIV capsid, a protein shell that protects the viral genome. To make the conical-shaped capsid, HIV CA proteins are organized
H. Tamura et al.
Mikrochim. Acta II, 287-287 (1983)
Kimura, K., et al.
J. Electroanal. Chem. Interfac. Electrochem., 95, 91-91 (1979)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service