Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55206

Supelco

4-Hydroxyphenylacetic acid

analytical standard

Synonym(s):

p-Hydroxyphenyl acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein/REAXYS Number:
1448766
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

assay

≥97.5% (GC)

shelf life

limited shelf life, expiry date on the label

mp

148-151 °C (lit.)

application(s)

clinical testing

format

neat

SMILES string

OC(=O)Cc1ccc(O)cc1

InChI

1S/C8H8O3/c9-7-3-1-6(2-4-7)5-8(10)11/h1-4,9H,5H2,(H,10,11)

InChI key

XQXPVVBIMDBYFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Hydroxyphenylacetic acid, a hydroxyphenylacetic acid metabolite, is reported to be produced via the bioactivation of kaempferol by intestinal microflora. It belongs to the class of dietary plant phenolic compounds, with a broad-spectrum of biological properties such as modulation of enzymatic activity, metal chelation, free radical scavenging, prevention of several human diseases, mainly atherosclerosis and cancer.

Application

4-Hydroxyphenylacetic acid may be used as an analytical reference standard for the quantification of the analyte in beer samples using high-performance liquid chromatography (HPLC) coupled to an electrochemical coulometric detector (ECD). It may also be used as an analytical standard for the quantification of the analyte in goat rumen fluid using HPLC with UV detection.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative determination of aromatic amino acids and related compounds in rumen fluid by high-performance liquid chromatography
Khan IR, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 710(1-2), 17-25 (1998)
Determination of free and bound phenolic acids in beer
Nardini M and Ghiselli A
Food Chemistry, 84(1), 137-143 (2004)
Cica Vissiennon et al.
The Journal of nutritional biochemistry, 23(7), 733-740 (2011-08-16)
Several in vivo and in vitro studies have confirmed that flavonols are metabolized by the intestinal microflora to their corresponding hydroxyphenylacetic acids. In this article, a comparison of the anxiolytic activity of the flavonols kaempferol, quercetin and myricetin in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service