Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55085

Sigma-Aldrich

8-Hydroxy-2-quinolinecarboxaldehyde 8-quinolylhydrazone

purum, ≥97.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14N4O
CAS Number:
Molecular Weight:
314.34
Beilstein/REAXYS Number:
491761
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

purum

assay

≥97.0% (HPLC)

mp

205-210 °C

SMILES string

Oc1cccc2ccc(\C=N\Nc3cccc4cccnc34)nc12

InChI

1S/C19H14N4O/c24-17-8-2-5-14-9-10-15(22-19(14)17)12-21-23-16-7-1-4-13-6-3-11-20-18(13)16/h1-12,23-24H/b21-12+

InChI key

MVRYUWHNRSHURE-CIAFOILYSA-N

Other Notes

Reagent for the sensitive determination of calcium[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E.A. Bozhevol'nov et al.
Zh. Anal. Khim., 24, 531-531 (1969)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service