Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50243

Supelco

Polylactic acid Standard 78′000

certified reference material (acc. to BAM), certified by BAM, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Polylactic acid, Poly(2-hydroxypropionic acid)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107

grade

certified reference material (acc. to BAM)

agency

certified by the BSC

quality

certified by BAM

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

gel permeation chromatography (GPC): suitable

format

neat

storage temp.

2-8°C

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)

InChI key

JVTAAEKCZFNVCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Exact molecular weight Mw, expiry date and detailed informaton certification can be found on the certificate and certification report, included in each package.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole Y C Yu et al.
European cells & materials, 25, 190-203 (2013-02-28)
We describe two studies encompassing the iterative refinement of a polymer-based rhBMP-2 delivery system for bone tissue engineering. Firstly, we compared the bone-forming capacity of porous poly(D,L-lactic-co-glycolic acid) (PLGA) scaffolds produced by thermally induced phase separation (TIPS) with non-porous solvent
Paul S Rosen et al.
Compendium of continuing education in dentistry (Jamesburg, N.J. : 1995), 34(1), 34-38 (2013-04-05)
This retrospective case series reports on the use of a polylactic acid barrier that was left exposed in the process of socket preparation for the placement of dental implants. A retrospective chart review found 43 patients with 48 extraction sockets
R M Felfel et al.
Materials science & engineering. C, Materials for biological applications, 33(4), 1914-1924 (2013-03-19)
Fibre reinforced composites have recently received much attention as potential bone fracture fixation applications. Bioresorbable composites based on poly lactic acid (PLA) and phosphate based glass fibre were investigated according to ion release, degradation, biocompatibility and mechanical retention profiles. The
David Goldberg et al.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 39(6), 915-922 (2013-03-08)
Injectable poly-L-lactic acid (PLLA) is a synthetic polymer indicated for the correction of facial wrinkles and folds. Animal studies have shown that implantation of PLLA stimulates collagen synthesis; human studies have been limited. To investigate human tissue response to injectable
Nomaan M Rezayee et al.
Dalton transactions (Cambridge, England : 2003), 42(15), 5573-5586 (2013-02-26)
A series of NNO ketoimines bearing trifluoromethyl substituents were synthesized from the Schiff base condensation of 1,3-diketones (1,1,1-trifluoro-5,5-dimethyl-2,4-hexanedione, 4,4,4-trifluoro-1-phenyl-1,3-butanedione, and 1,1,1-trifluoro-2,4-pentanedione, 1,1,1,5,5,5-hexafluoroacetylacetone) and 8-aminoquinoline or 8-amino-2-methylquinoline and isolated in 40-70% yield. The ketoimines were combined with zinc bis-(trimethylsilyl)amide to prepare

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service