Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

49990

Sigma-Aldrich

sn-Glycerol 3-phosphate bis(cyclohexylammonium) salt

≥94.0% (GC)

Synonym(s):

L-α-Glycerol phosphate bis(cyclohexylammonium) salt, L-α-Glycerophosphate bis(cyclohexylammonium) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H9O6P · 2C6H13N
CAS Number:
Molecular Weight:
370.42
Beilstein/REAXYS Number:
6831546
EC Number:
MDL number:
UNSPSC Code:
12352201

assay

≥94.0% (GC)

storage temp.

−20°C

SMILES string

NC1CCCCC1.NC2CCCCC2.OC[C@@H](O)COP(O)(O)=O

InChI

1S/2C6H13N.C3H9O6P/c2*7-6-4-2-1-3-5-6;4-1-3(5)2-9-10(6,7)8/h2*6H,1-5,7H2;3-5H,1-2H2,(H2,6,7,8)/t;;3-/m..1/s1

InChI key

FWIDSLORBURWJY-CFMHHUGTSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher J Law et al.
Journal of molecular biology, 378(4), 828-839 (2008-04-09)
Active transport of substrates across cytoplasmic membranes is of great physiological, medical and pharmaceutical importance. The glycerol-3-phosphate (G3P) transporter (GlpT) of the E. coli inner membrane is a secondary active antiporter from the ubiquitous major facilitator superfamily that couples the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service