Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45354

Sigma-Aldrich

[2-(3,4-Epoxycyclohexyl)ethyl]trimethoxysilane

purum, ≥97.0% (GC)

Synonym(s):

2-(7-Oxabicyclo[4.1.0]hept-3-yl)ethyl-trimethoxysilane, 3-(2-Trimethoxysilylethyl)cyclohexene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H22O4Si
CAS Number:
Molecular Weight:
246.38
Beilstein/REAXYS Number:
8980119
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

purum

assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.451 (lit.)
n20/D 1.451

bp

310 °C (lit.)

density

1.065 g/mL at 25 °C (lit.)

SMILES string

CO[Si](CCC1CCC2OC2C1)(OC)OC

InChI

1S/C11H22O4Si/c1-12-16(13-2,14-3)7-6-9-4-5-10-11(8-9)15-10/h9-11H,4-8H2,1-3H3

InChI key

DQZNLOXENNXVAD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service