Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44715

Sigma-Aldrich

Fast Green FCF

for microscopy

Synonym(s):

Food green 3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C37H34N2O10S3Na2
CAS Number:
Molecular Weight:
808.85
Colour Index Number:
42053
Beilstein/REAXYS Number:
5718212
EC Number:
MDL number:
UNSPSC Code:
12171500

grade

for microscopy

mp

290 °C (dec.) (lit.)

SMILES string

[Na+].[Na+].CCN(Cc1cccc(c1)S([O-])(=O)=O)c2ccc(cc2)C(=C3/C=CC(\C=C3)=[N+](\CC)Cc4cccc(c4)S([O-])(=O)=O)\c5ccc(O)cc5S([O-])(=O)=O

InChI

1S/C37H36N2O10S3.2Na/c1-3-38(24-26-7-5-9-33(21-26)50(41,42)43)30-15-11-28(12-16-30)37(35-20-19-32(40)23-36(35)52(47,48)49)29-13-17-31(18-14-29)39(4-2)25-27-8-6-10-34(22-27)51(44,45)46;;/h5-23H,3-4,24-25H2,1-2H3,(H3,41,42,43,44,45,46,47,48,49);;/q;2*+1/p-2

InChI key

RZSYLLSAWYUBPE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Fast Green FCF dye is used for protein staining in IEF, native PAGE and SDS-PAGE. Fast Green staining is linear over a wider range of protein concentrations than Brilliant Blue R. After electrophoresis, fixing the proteins in the gel is recommended for maximum sensitivity. The gel can then be stained for 2 hr with 0.1% Fast Green FCF in either 30% (v/v) ethanol, 10% (v/v) acetic acid or in 7% acetic acid. The gel can be destained with either 30% (v/v) ethanol, 10% (v/v) acetic acid, or 7% acetic acid. The stained gel can be scanned at 625 nm. The detection sensitivity is approx. 30% that with Brilliant Blue R.

Other Notes

Reagent for staining proteins after PAGE[1]; Especially suitable in isoelectric focusing (does not bind to ampholytes)[2]

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Staining protein in isoelectric focusing gels with fast green.
R E Allen et al.
Analytical biochemistry, 104(2), 494-498 (1980-05-15)
Studies and critique of Amido Black 10B, Coomassie Blue R, and Fast Green FCF as stains for proteins after polyacrylamide gel electrophoresis.
C M Wilson
Analytical biochemistry, 96(2), 263-278 (1979-07-15)
Sumiko Tsuji et al.
Journal of chromatography. A, 1101(1-2), 214-221 (2005-10-26)
The existence of five of the six expected isomers in commercial Fast Green FCF (G3: Food Green No. 3, FD&C Green No. 3, CAS No. 2353-45-9, C.I. No. 42053), the main product of which is m,m-G3 and the sub-products of
A V Grigor'eva et al.
Tsitologiia, 27(2), 191-195 (1985-02-01)
A cytophotometrical study was made of Fast Green FCF binding by nuclear non-histone proteins of the rat superior ganglion neurons at pH 2.6 (1 day, 1 week, 1 and 5 months, and 2.5 years after birth, a. b.). An increased
R Moss et al.
Stain technology, 57(1), 1-4 (1982-01-01)
It has been shown that gluten powder stained for one minute and forty-five seconds in a 0.192% solution of fast green FCF and then washed repeatedly with distilled water can be detected both by visible light and fluorescence microscopy when

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service