Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43959

Supelco

Methyl stearidonate

≥97.0% (GC)

Synonym(s):

cis-6,9,12,15-Octadecatetraenoic acid methyl ester, Stearidonic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30O2
CAS Number:
Molecular Weight:
290.44
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

assay

≥97.0% (GC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

functional group

ester

shipped in

ambient

storage temp.

−20°C

SMILES string

CC\C=C/C\C=C/C\C=C/C\C=C/CCCCC(=O)OC

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J González-Fernández et al.
Biochimie, 139, 107-114 (2017-06-10)
Important health benefits have been attributed to monoacylglycerols (MAGs) due to their various physiological functions, owing to which they become candidates for use as functional foods in order to prevent the onset of certain diseases such as colon cancer. In
Miguel Urrestarazu et al.
PloS one, 14(2), e0207106-e0207106 (2019-02-20)
Borage is a well-known plant of great importance in human nutrition and health. Expanding knowledge of particular plants that have anti-cancer products is a global concern. There is substantial information regarding the benefits, presence and extraction of gamma linolenic acid
María José González-Fernández et al.
Nutrients, 11(12) (2019-12-11)
Colorectal cancer (CRC) is one of the most common and mortal types of cancer. There is increasing evidence that some polyunsaturated fatty acids (PUFAs) exercise specific inhibitory actions on cancer cells through different mechanisms, as a previous study on CRC
Robert Bodkowski et al.
Animals : an open access journal from MDPI, 10(10) (2020-10-15)
The study aimed to investigate the effect of supplementation of fish oil after the process of low-temperature crystallization (LTC-FO) enriched with long-chain polyunsaturated fatty acids (LC-PUFAs) on cow milk parameters. The experiment was carried out on 24 Polish Holstein Friesian
Ana Penko et al.
Food chemistry, 168, 372-382 (2014-08-31)
The oxidation processes were studied in chicken patties, enriched with n-3 fatty acids, after 8days of storage at 4°C, under different aerobic conditions, and following heat treatment. Significant effects were seen on lipid and cholesterol oxidation and the sensory qualities

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service