Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42070

Sigma-Aldrich

2,4-Dinitro-5-fluoroaniline

≥98.0% (HPLC)

Synonym(s):

Bergmann’s Reagent, DNFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H2(NO2)2NH2
CAS Number:
Molecular Weight:
201.11
Beilstein/REAXYS Number:
2280981
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

≥98.0% (HPLC)

mp

185-190 °C (lit.)
185-190 °C

SMILES string

Nc1cc(F)c(cc1[N+]([O-])=O)[N+]([O-])=O

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for labeling free amino groups in proteins before hydrolysis. DNFA is reported to have several advantages over the Sanger reagent, dinitrofluorobenzene (Cat. No. 42080)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E.D. Bergmann et al.
The Journal of Organic Chemistry, 26, 1480-1480 (1961)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service