Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41810

Sigma-Aldrich

1,2-Dimyristoyl-3-palmitoyl-rac-glycerol

≥99.0% (TLC)

Synonym(s):

1,2-Ditetradecanoyl-3-hexadecanoyl-rac-glycerol, rac-Glycerol 1,2-dimyristate 3-palmitate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H90O6
CAS Number:
Molecular Weight:
751.21
Beilstein/REAXYS Number:
1811058
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

assay

≥99.0% (TLC)

solubility

chloroform: 50 mg/mL, clear, colorless

suitability

in accordance for fatty acid composition

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCC

InChI

1S/C47H90O6/c1-4-7-10-13-16-19-22-23-26-28-31-34-37-40-46(49)52-43-44(53-47(50)41-38-35-32-29-25-21-18-15-12-9-6-3)42-51-45(48)39-36-33-30-27-24-20-17-14-11-8-5-2/h44H,4-43H2,1-3H3

InChI key

AMXRERGJDVUTMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service