Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41713

Sigma-Aldrich

N,N-Dimethyltridecylamine N-oxide

≥97.0% (NT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12N(O)(CH3)2
CAS Number:
Molecular Weight:
243.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

description

non-ionic

assay

≥97.0% (NT)

impurities

≤0.03% peroxides (as H2O2)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤5 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCC[N+](C)(C)[O-]


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service