40893
Peptone Water, phosphate-buffered, Vegitone
suitable for microbiology, NutriSelect® Plus
Synonym(s):
BPW Vegitone, Buffered Peptone Water Vegitone, Vegitone Peptone Water, phosphate-buffered
About This Item
form
powder
Quality Level
shelf life
limited shelf life, expiry date on the label
composition
disodium phosphate, 3.5 g/L
monopotassium phosphate, 1.5 g/L
proteose pepetone (vegetable), 10.0 g/L
sodium chloride, 5.0 g/L
manufacturer/tradename
NutriSelect® Plus
technique(s)
microbiological culture: suitable
final pH
7.2±0.2 (25 °C)
application(s)
agriculture
environmental
food and beverages
sample preparation
microbiology
suitability
Providencia spp.
nonselective for Citrobacter spp.
nonselective for Escherichia coli
nonselective for Klebsiella spp.
nonselective for Proteus spp.
nonselective for Pseudomonas spp.
nonselective for Salmonella spp.
nonselective for Shigella spp.
nonselective for Staphylococcus spp.
nonselective for Streptococcus spp.
nonselective for Yersinia spp.
nonselective for bacteria (General Media)
nonselective for coliforms
nonselective for enterobacteriaceae
Application
Legal Information
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
An article concerning the detection, identification, differentiation, and cultivation of Pseudomonas species.
Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service