40865
2,3-Dimethyl-6-nitroaniline
technical, ≥95% (CHN)
Synonym(s):
6-Nitro-2,3-xylidine
About This Item
grade
technical
assay
≥95% (CHN)
mp
117-120 °C (lit.)
117-120 °C
SMILES string
Cc1ccc(c(N)c1C)[N+]([O-])=O
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service